missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl-Beta-D-glucuronide, 1mg/ml in Methanol, Lipomed
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Description
Ethyl-Beta-D-glucuronide, 1mg/ml in Methanol
Specifications
Specifications
| Chemical Name or Material | Ethyl-beta-D-glucuronide |
| CAS | 17685-04-0 |
| Analyte or Component Names | Ethyl-beta-D-glucuronide |
| Shipping Condition | Ice Pk. (-20°C) |
| Molecular Formula | C8 H14 O7 |
| Concentration | 1 mg/mL |
| Synonym | β-D-Glucopyranosiduronic acid, ethyl (9 CI, ACI), Ethyl β-D-glucopyranosiduronic acid (ACI), Glucopyranosiduronic acid, ethyl, β-D- (6 CI, 8 CI), BA 2829, Ethyl glucuronide, Ethyl β-D-glucosiduronic acid, Ethyl β-D-glucuronide |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCO[C@@H]1 O[C@@H]([C@@H](O)[C@H](O)[C@H]1 O)C(=O)O |
| IUPAC Name | (2 S,3 S,4 S,5 R,6 R)-6-ethoxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering