missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Beta-Carboline-3-carboxylate (Beta-CCE), TRC

Product Code. 30382165
Change view
Click to view available options
Quantity:
100 mg
250 mg
Unit Size:
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382165 100 mg -
30369396 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30382165 Supplier Toronto Research Chemicals Supplier No. TRCE901503100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74214-62-3
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
IUPAC Name ethyl 9H-pyrido[3,4-b]indole-3-carboxylate
SMILES CCOC(=O)c1cc2c(cn1)[nH]c3ccccc23

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H12N2O2/c1-2-18-14(17)12-7-10-9-5-3-4-6-11(9)16-13(10)8-15-12/h3-8,16H,2H2,1H3
Formula Weight 240.0899
Chemical Name or Material Ethyl beta-Carboline-3-carboxylate (beta-CCE)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.