missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzoylacetate, 90+%

Product Code. 11381797
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11381797 1000 g 1kg
11361797 50 g 50g
11371797 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11381797 Supplier Thermo Scientific Alfa Aesar Supplier No. L05030.0B
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-02-0 | C11H12O3 | 192.214 g/mol

Ethyl benzoylacetate is used as a flavoring agent and an intermediate in organic synthesis. Further, it is used in the preparation of triazipinones, iodonium ylides, ethyl 2-fluoro-2-benzolyacetate and benzo[c]furan-4,7-diones. It is involved in the Clisen condensation reaction with malononitrile to prepare 2-cyano-5-phenyl-3,5-dioxopentanonitrile.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl benzoylacetate is used as a flavoring agent and an intermediate in organic synthesis. Further, it is used in the preparation of triazipinones, iodonium ylides, ethyl 2-fluoro-2-benzolyacetate and benzo[c]furan-4,7-diones. It is involved in the Clisen condensation reaction with malononitrile to prepare 2-cyano-5-phenyl-3,5-dioxopentanonitrile.

Solubility
Miscible with ethanol and ether. Immiscible with water.

Notes
Air and light sensitive. Incompatible with acids, bases, oxidizing agents and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-02-0
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00009196
InChI Key GKKZMYDNDDMXSE-UHFFFAOYSA-N
Synonym ethyl benzoylacetate, benzoylacetic acid ethyl ester, ethyl benzoyl acetate, acetic acid, benzoyl-, ethyl ester, ethyl beta-oxobenzenepropanoate, ethylbenzoylacetate, benzoylacetic acid, ethyl ester, ethyl 3-phenyl-3-oxopropanoate, benzenepropanoic acid, beta-oxo-, ethyl ester, ethyl 3-oxo-3-phenylpropionate
PubChem CID 7170
IUPAC Name ethyl 3-oxo-3-phenylpropanoate
SMILES CCOC(=O)CC(=O)C1=CC=CC=C1

Specifications

Density 1.113
Boiling Point 266°C to 270°C
Flash Point 140°C (284°F)
Refractive Index 1.529
Quantity 1000 g
Beilstein 389944
Merck Index 14,3767
Solubility Information Miscible with ethanol and ether. Immiscible with water.
Formula Weight 192.21
Percent Purity 96%
Chemical Name or Material Ethyl benzoylacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.