missing translation for 'onlineSavingsMsg'
Learn More

Ethyl benzothiazole-2-carboxylate, 98%

Product Code. 15425677
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425677 5 g 5g
15435677 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425677 Supplier Thermo Scientific Alfa Aesar Supplier No. H32904.06
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 32137-76-1 | C10H9NO2S | 207.247 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32137-76-1
Molecular Formula C10H9NO2S
Molecular Weight (g/mol) 207.247
MDL Number MFCD00848360
InChI Key VLQLCEXNNGQELL-UHFFFAOYSA-N
Synonym ethyl benzo d thiazole-2-carboxylate, ethyl benzothiazole-2-carboxylate, benzothiazole-2-carboxylic acid ethyl ester, 2-benzothiazolecarboxylic acid, ethyl ester, ethyl 2-benzothiazolecarboxylate, ethyl1,3-benzothiazole-2-carboxylate, zlchem 880, pubchem23783, acmc-209hra, ksc490c5n
PubChem CID 640708
IUPAC Name ethyl 1,3-benzothiazole-2-carboxylate
SMILES CCOC(=O)C1=NC2=CC=CC=C2S1

Specifications

Melting Point 68°C to 72°C
Quantity 5 g
Formula Weight 207.25
Percent Purity 98%
Chemical Name or Material Ethyl benzothiazole-2-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.