missing translation for 'onlineSavingsMsg'
Learn More

(Ethyl benzoate)tricarbonylchromium, 98%, Thermo Scientific™

Product Code. 11379818
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11379818 5 g 5g
11389818 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379818 Supplier Thermo Scientific Alfa Aesar Supplier No. 044283.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Hydrogenation catalyst

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32874-26-3
Molecular Formula C12H10CrO5
Molecular Weight (g/mol) 286.203
MDL Number MFCD02683559
InChI Key SWSAJUVURPSGIG-UHFFFAOYSA-N
Synonym ethyl benzoate tricarbonylchromium, ethyl benzoate tricarbonylchromium 0, tris carbon monoxide ethyl benzoate chromium
PubChem CID 13222494
IUPAC Name carbon monoxide;chromium;ethyl benzoate
SMILES CCOC(=O)C1=CC=CC=C1.[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr]

Specifications

Melting Point 54°C to 59°C
Color Orange
Quantity 5 g
UN Number UN3466
Sensitivity Air and moisture sensitive
Formula Weight 286.2
Percent Purity 98%
Physical Form Powder
Chemical Name or Material (Ethyl benzoate)tricarbonylchromium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.