missing translation for 'onlineSavingsMsg'
Learn More

Ethyl azidoacetate, 25% solution in toluene, Thermo Scientific Chemicals

Catalog Number 13408628
Click to view available options
25mL, Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 637-81-0, 108-88-3
Density 0.9200g/mL
Quantity 25 mL
Flash Point 7°C
Packaging Glass Bottle
Specific Gravity 0.92
Molecular Formula C4H7N3O2
Synonym ethyl azidoacetate, azidoacetic acid ethyl ester, acetic acid, azido-, ethyl ester, azido-acetic acid ethyl ester, ccris 3368, acetic acid, 2-azido-,ethyl ester, ethyl azido-acetate, ethyl2-azidoacetate, ethyl-2-azidoacetate, ethylazidoacetatesolution
InChI Key HVJJYOAPXBPQQV-UHFFFAOYSA-N
SMILES CCOC(=O)CN=[N+]=[N-]
IUPAC Name ethyl 2-azidoacetate
Molecular Weight (g/mol) 129.12
PubChem CID 69480
Formula Weight 129.12
Chemical Name or Material Ethyl azidoacetate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethyl azidoacetate, 25% solution in toluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'