missing translation for 'onlineSavingsMsg'
Learn More

Ethyl alpha,4-dibromophenylacetate, 97+%

Product Code. 11397579
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397579 5 g 5g
11307589 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397579 Supplier Thermo Scientific Alfa Aesar Supplier No. H28093.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77143-76-1 | C10H10Br2O2 | 321.996 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77143-76-1
Molecular Formula C10H10Br2O2
Molecular Weight (g/mol) 321.996
MDL Number MFCD09953497
InChI Key PPRAWWBOUOBAON-UHFFFAOYSA-N
Synonym ethyl 2-bromo-2-4-bromophenyl acetate, ethyl 2-bromo-4-bromophenyl acetate, ethyl bromo 4-bromophenyl acetate, alpha,4-dibromophenylacetic acid ethyl ester, ethyl alpha,4-dibromophenylacetate, ethyl2-bromo-2-4-bromophenyl acetate, ethyl 2-bromanyl-2-4-bromophenyl ethanoate, benzeneacetic acid, ?,4-dibromo-, ethyl ester, benzeneacetic acid, a,4-dibromo-, ethyl ester
PubChem CID 12754876
IUPAC Name ethyl 2-bromo-2-(4-bromophenyl)acetate
SMILES CCOC(=O)C(C1=CC=C(C=C1)Br)Br

Specifications

Density 1.642
Boiling Point 140°C (5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.571
Quantity 5 g
UN Number UN3265
Solubility Information Slightly soluble in water.
Formula Weight 321.99
Percent Purity ≥97%
Chemical Name or Material Ethyl alpha,4-dibromophenylacetate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.