missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Alcohol, Pure 190 proof, Honeywell™ Riedel-de-Haën™

Product Code. 15667850
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15667850 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15667850 Supplier Honeywell Riedel-de Haën Supplier No. E7148500ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point -131°C
CAS 64-17-5
Density 0.816g/mL at 25°C
Quantity 500 mL
Flash Point 14°C (57.2°F)
Molecular Formula C2H6O
Linear Formula CH3CH2OH
MDL Number MFCD00003568
UN Number UN1170
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
Boiling Point 36°C to 37°C
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Concentration or Composition (by Analyte or Components) 190 proof
Chemical Name or Material Ethyl alcohol, Pure, For molecular biology
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.