missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acrylate (Stabilized with 4-methoxyphenol), TRC

Product Code. 30383623
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383623 25 g
30380979 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383623 Supplier Toronto Research Chemicals Supplier No. TRCE89730025G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 140-88-5
Molecular Formula C5 H8 O2
Molecular Weight (g/mol) 100.12
Synonym Acrylic acid ethyl ester (6CI,8CI), 2-Propenoic acid ethyl ester, Ethyl 2-propenoate, Ethyl acrylate, Ethyl acrylic ester, Ethyl propenoate, NSC 8263
IUPAC Name ethyl prop-2-enoate
SMILES CCOC(=O)C=C

Specifications

Quantity 25 g
InChI Formula InChI=1S/C5H8O2/c1-3-5(6)7-4-2/h3H,1,4H2,2H3
Formula Weight 100.0524
Chemical Name or Material Acrylic acid-ethyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.