missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetolactate, TRC

Product Code. 30380500
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380500 25 mg
30384316 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380500 Supplier Toronto Research Chemicals Supplier No. TRCE89619025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16508-89-7
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
Synonym Ethyl 2-Hydroxy-2-methyl-3-oxobutanoate, 2-Hydroxy-2-methyl-3-oxo-butanoic Acid Ethyl Ester, 2-Hydroxy-2-methyl-acetoacetic Acid Ethyl Ester
SMILES CCOC(=O)C(C)(O)C(=O)C

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C7H12O4/c1-3-11-7(10)6(9)4-5(2)8/h6,9H,3-4H2,1-2H3
Formula Weight 160.0736
Chemical Name or Material Ethyl Acetolactate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.