missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetoacetate, TRC

Product Code. 30376789
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30376789 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376789 Supplier Toronto Research Chemicals Supplier No. TRCE89954550G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl Acetoacetate
CAS 141-97-9
Molecular Formula C6H10O3
Quantity 50 g
Synonym Ethyl Acetoacetate-1,2,3,4, 1-Ethoxybutane-1,3-dione, 3-Oxobutanoic Acid Ethyl Ester, 3-Oxobutyric Acid Ethyl Ester, Acetylacetic Acid Ethyl Ester, EAA, Ethyl 2-Acetoacetate, Ethyl 2-Methyl-3-oxopropionate, Ethyl 3-Ketobutyrate, Ethyl 3-Oxobutanoate, Ethyl 3-Oxobutyrate, Ethyl Acetoacetate, Ethyl Acetonecarboxylate, Ethyl Acetylacetate, NSC 37390, NSC 8657
InChI Formula InChI=1S/C6H10O3/c1-3-9-6(8)4-5(2)7/h3-4H2,1-2H3
SMILES CCOC(=O)CC(=O)C
IUPAC Name ethyl 3-oxobutanoate
Molecular Weight (g/mol) 130.14
Formula Weight 130.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.