missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetoacetate propylene glycol ketal, 97%, Thermo Scientific™

Product Code. p-7046226
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11353117

Brand: Thermo Scientific Alfa Aesar L13514.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6290-17-1
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
MDL Number MFCD00151819
InChI Key GSIXJEIRJVOUFB-UHFFFAOYSA-N
Synonym ethyl 2-2,4-dimethyl-1,3-dioxolan-2-yl acetate, 1,3-dioxolane-2-acetic acid, 2,4-dimethyl-, ethyl ester, ethyl acetoacetate propylene glycol ketal, fraistone, ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate, ethyl dimethyl dioxolane acetate, acetoacetic acid, ethyl ester, 1,2-propylene ketal, cis-and trans-ethyl 2,4-dimethyl-1,3-dioxolane-2-acetate, ethyl 2,4-dimethyl-1,3-dioxolan-2-yl acetate, 2,4-dimethyl-1,3-dioxolane-2-acetic acid ethyl ester
PubChem CID 95392
IUPAC Name ethyl 2-(2,4-dimethyl-1,3-dioxolan-2-yl)acetate
SMILES CCOC(=O)CC1(OCC(O1)C)C

Specifications

Boiling Point 90°C (10 mmHg)
Flash Point 65°C (149°F)
Refractive Index 1.428
Quantity 25 g
Beilstein 138927
Formula Weight 188.23
Percent Purity 97%
Chemical Name or Material Ethyl acetoacetate propylene glycol ketal
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.