missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetoacetate, 99+%, extra pure

Product Code. 10127743 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127743 2.5 kg Glass bottle 2.5kg
10452031 100 g Glass bottle 100g
10380734 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127743 Supplier Thermo Scientific Acros Supplier No. 220400025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-97-9 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-97-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00009199
InChI Key XYIBRDXRRQCHLP-UHFFFAOYSA-N
Synonym ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
PubChem CID 8868
ChEBI CHEBI:4893
IUPAC Name ethyl 3-oxobutanoate
SMILES CCOC(=O)CC(=O)C

Specifications

Melting Point -45°C
Density 1.0300g/mL
Boiling Point 180°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCH2CO2C2H5
Refractive Index 1.4180 to 1.42
Quantity 2.5 kg
Beilstein 03, 632
Fieser 12,222
Merck Index 15, 3812
Specific Gravity 1.03
Solubility Information Solubility in water: 116g/L (20°C). Other solubilities: soluble in most organic solvent: diethyl ether,,benzene,ethanol,ehyl acetate,chloroform,,acetone
Viscosity 1.696 mPa.s (20°C)
Formula Weight 130.14
Percent Purity 99+%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Ethyl acetoacetate, p.a.
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.