missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetoacetate, 99+%

Product Code. 11404234
Change view
Click to view available options
Quantity:
10,000 g
250 g
2500 g
500 g
Unit Size:
10000g
2500g
250g
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11404234 2500 g 2500g
11458940 250 g 250g
11494224 500 g 500g
11414234 10,000 g 10000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11404234 Supplier Thermo Scientific Alfa Aesar Supplier No. A12544.0E
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 141-97-9 | C6H10O3 | 130.143 g/mol

Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl acetoacetate is used as an intermediate in organic synthesis and as a co-promoter for unsaturated polyester resins. It is widely used in the production of dyes, inks, perfumes, plastics and flavoring agents. It is an important starting material for the syntheses of alpha-substituted acetoacetic esters and cyclic compounds like pyrazole, pyrimidine and coumarin derivatives. It acts as an intermediate in the synthesis of vitamins and pharmaceuticals. It finds application as a formaldehyde scavenger.

Solubility
Miscible with water, benzene, chloroform, ether and acetone.

Notes
Incompatible with acids, bases, oxidizing agents, alkali metals and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-97-9
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.143
MDL Number MFCD00009199
InChI Key XYIBRDXRRQCHLP-UHFFFAOYSA-N
Synonym ethyl acetoacetate, ethyl acetylacetate, ethyl 3-oxobutyrate, diacetic ether, butanoic acid, 3-oxo-, ethyl ester, ethyl acetylacetonate, acetoacetic acid, ethyl ester, 3-oxobutanoic acid ethyl ester, 3-oxo-butyric acid ethyl ester, active acetylacetate
PubChem CID 8868
ChEBI CHEBI:4893
IUPAC Name ethyl 3-oxobutanoate
SMILES CCOC(=O)CC(=O)C

Specifications

Melting Point -43°C
Density 1.025
Boiling Point 180°C to 181°C
Flash Point 84°C (183°F)
Assay Percent Range ≥99%
Refractive Index 1.419
Quantity 2500 g
Beilstein 385838
Merck Index 14,3758
Solubility Information Miscible with water,benzene,chloroform,ether and acetone.
Formula Weight 130.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Ethyl Acetoacetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.