missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetimidate Hydrochloride (>90%), TRC

Product Code. 30377413
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30377413 10 g 10g
30371855 5 g -
30373131 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30377413 Supplier Toronto Research Chemicals Supplier No. TRCE89882810G
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2208-07-3
Molecular Formula C4H9NO . HCl
Molecular Weight (g/mol) 123.58
Synonym Ethanimidic acid, ethyl ester, hydrochloride (1:1) (ACI), Acetimidic acid, ethyl ester, hydrochloride (6CI, 7CI, 8CI), Ethanimidic acid, ethyl ester, hydrochloride (9CI), Ethyl acetimidate hydrochloride, Ethyl acetoamidate hydrochloride, Ethyl acetocarboximidate hydrochloride, Ethyl acetoimidate hydrochloride, Ethyl ethanimidate hydrochloride, Ethyl ethanimidoate hydrochloride, Ethyl iminoacetate hydrochloride, Acetimidic acid ethyl ester hydrochloride, Ethanimidic acid ethyl ester hydrochloride
IUPAC Name ethyl ethanimidate;hydrochloride
SMILES Cl.CCOC(=N)C

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H9NO.ClH/c1-3-6-4(2)5;/h5H,3H2,1-2H3;1H
Formula Weight 123.0451
Chemical Name or Material Ethyl Acetimidate Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.