missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, Puriss., Solstice

Product Code. 17258316
Change view
Click to view available options
Quantity:
1000 L
Unit Size:
1000 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17258316 1000 L 1000 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17258316 Supplier Honeywell Riedel-de Haën Supplier No. 27227S1000L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point -84°C
CAS 141-78-6
Color Colorless
Quantity 1000 L
Packaging Returnable Stainless Steel IBC
Molecular Formula C4H8O2
Synonym EtOAc
Boiling Point 77°C
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES CCOC(C)=O
IUPAC Name ethyl acetate
Molecular Weight (g/mol) 88.11
Percent Purity ≥99.5% (GC)
Grade puriss.
Chemical Name or Material Ethyl acetate
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Ethyl acetate
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.