missing translation for 'onlineSavingsMsg'
Learn More

Ethyl Acetate, BAKER ANALYZED Pesticide Reagent, J.T. Baker™

Product Code. 15232178 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15232178 2.5 L Glass 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15232178 Supplier JT Baker Supplier No. 5277.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specially developed for use in evaluation of pesticide and insecticide residues in food, feed, water or soil samples. Ethyl Acetate, BAKER ANALYZED Pesticide Reagent, J.T. Baker™ is suitable for all analysis of common pesticides.

J.T. Baker BAKER ANALYZED Pesticide Reagent Solvents:

  • Suitable for analysis of most common pesticides, such as 2-chlorobiphenyl , Heptachlor, Aldrin, Parathion, Dieldrin, Endrin, DDT, Dioxine, etc.
  • Specified to the required low residue levels (5 ppm) for use in research and quality control laboratories
  • Ideal for gas chromatography applications, testing for ECD and/or FID sensitive impurities, organic residues, and residue after evaporation

Specifications

Melting Point -83.57°C
CAS 141-78-6
Color Colorless
Acidity Max. 0.0005meq/g
Density 0.9g/cm3 at 20°C
Quantity 2.5 L
Explosion Limit 2.1 - 11.5 Vol %
Flash Point −1°C
Ignition Point 460°C
Odor Fruit-like
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 5 ppm
Molecular Formula C4H8O2
Linear Formula CH3-COO-CH2-CH3
Viscosity 0.44 mPas at 20°C
Water Max. 0.05%
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
Boiling Point 76°C to 78°C
Solubility Information 79g/L in water
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES CCOC(=O)C
IUPAC Name ethyl acetate
Molecular Weight (g/mol) 88.106
PubChem CID 8857
ChEBI CHEBI:27750
Formula Weight 88.11g/mol
Percent Purity Min. 99.8%
Grade Pesticide
Chemical Name or Material Ethyl Acetate
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.