missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, ACS, 99.5+%, Thermo Scientific Chemicals

Product Code. 11317458
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
4 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Ethyl acetate is widely used as a solvent in cleaning of the circuit board. It is most suited for HPLC instruments and spectrophotometers including the UV-visible spectrophotometer, the IR spectrometer and the mass spectrometer. In addition, it can also be used as a solvent in column chromatography and solvent extraction. It is also used in glues, nail polish removers, decaffeinating tea and coffee and in cigarettes. It is present in confectioneries, perfumes and fruits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.106
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
PubChem CID 8857
ChEBI CHEBI:27750
IUPAC Name ethyl acetate
SMILES CCOC(=O)C

Specifications

Melting Point -84°C
Color Colorless
Density 0.902 g/mL
Boiling Point 77°C
Quantity 20 L
Flash Point -3°C (26°F)
Odor Sweet
Physical Form Liquid
Refractive Index 1.372
Linear Formula CH3CO2CH2CH3
UN Number 1173
Beilstein 506104
Merck Index 14,3757
Solubility Information Miscible with ethanol,acetone,diethyl ether and benzene.
Formula Weight 88.11
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Ethyl Acetate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'