missing translation for 'onlineSavingsMsg'
Learn More

Ethyl acetate, Laboratory Reagent, ≥99.0% (GC), Solstice

Product Code. 15671550
Change view
Click to view available options
Quantity:
4 x 5 L
Packaging:
Aluminum Bottle
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15671550 4 x 5 L Aluminum Bottle Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15671550 Supplier Fluka Supplier No. 163714X5L
 Restricted Item
Request Bulk or Custom Formats
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Laboratory Reagent,≥99.0% (GC)

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Specifications

CAS 141-78-6
Density 0.902g/mL at 25°C
Quantity 4 x 5 L
Explosion Limit 2.2 to 11.5% (38°F)
Flash Point −3°C (26.6°F)
Ignition Point 801°F
Packaging Aluminum Bottle
Molecular Formula C4H8O2
Linear Formula CH3COOC2H5
Vapor Pressure 73mmHg (20°C)
MDL Number MFCD00009171
UN Number UN1173
Beilstein 506104
Synonym ethyl ethanoate, acetoxyethane, acetic acid ethyl ester, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, ethylacetate, acetidin, acetic ester
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES CCOC(=O)C
IUPAC Name ethyl acetate
Molecular Weight (g/mol) 88.106
PubChem CID 8857
ChEBI CHEBI:27750
Formula Weight 88.11g/mol
Percent Purity ≥99.0% (GC)
Grade Laboratory
Chemical Name or Material Ethyl Acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethyl acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.