missing translation for 'onlineSavingsMsg'
Learn More

Solstice Ethyl Acetate

Product Code. 15925470
Change view
Click to view available options
Quantity:
7 L
18 L
45 L
1000 L
Unit Size:
1000 L
18 L
45 L
7 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15925470 7 L 7 L
15627025 18 L 18 L
15915470 45 L 45 L
16625823 1000 L 1000 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15925470 Supplier Solstice Supplier No. 332117L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Ethyl Acetate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym EtOAc
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

pH No data available
Melting Point -84°C
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.901 g/cm3
Quantity 7 L
Flash Point 11.5 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula CH3COOC2H5
Vapor Pressure 97 hPa (20°C)
Viscosity No data available
UN Number UN1173
Beilstein 506104
Concentration ≥99.50%
Boiling Point 77°C (1.013 hPa)
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material Ethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.