missing translation for 'onlineSavingsMsg'
Learn More

Solstice Ethyl Acetate

Product Code. 15925440
Change view
Click to view available options
Quantity:
18 L
Unit Size:
18 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15925440 18 L 18 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15925440 Supplier Solstice Supplier No. 2722718L
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ethyl Acetate, Puriss., meets analytical specification of BP, NF, Ph. Eur., ≥99.5% (GC).

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals.
  • It is the leading brand offering High-Purity Solvents for dedicated applications.

Specifications

pH No data available
Melting Point -84°C
CAS 141-78-6
Color Colorless
Density 0.901 g/cm3
Quantity 18 L
Flash Point 11.5 %(V)
Packaging Returnable Stainless Steel Can
Physical Form Liquid
Product Line Puriss.
Molecular Formula C4H8O2
Linear Formula CH3COOC2H5
Vapor Pressure 97 hPa (20°C)
MDL Number MFCD00009171
UN Number UN1173
Beilstein 506104
Synonym EtOAc
Boiling Point 77°C (1.013 hPa)
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
SMILES CCOC(C)=O
IUPAC Name ethyl acetate
Molecular Weight (g/mol) 88.11
Percent Purity ≥99.50%
Grade GC
Chemical Name or Material Ethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.