missing translation for 'onlineSavingsMsg'
Learn More

Riedel-de Haën Ethyl Acetate

Product Code. 15915470
Click to view available options
Quantity:
7 L
18 L
45 L
1000 L
Unit Size:
1000 L
18 L
45 L
7 L
This item is not returnable. View return policy

Product Code. 15915470

Brand: Riedel-de Haën 3321145L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

Ethyl Acetate, Puriss. p.a., ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99.5% (GC).

  • ACS general use solvents.
  • Value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis.
  • Suitable for less demanding applications such as routine isocratic HPLC.
  • Variations and decrease issues traceable to solvent contamination.
  • All solvents are tested to meet ACS general use specifications.

Chemical Identifiers

CAS 141-78-6
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00009171
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym EtOAc
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

pH No data available
Melting Point -84°C
CAS Min % ≥90.00%
Color Colorless
CAS Max % ≤100.00%
Density 0.901 g/cm3
Boiling Point 77°C (1.013 hPa)
Quantity 45 L
Flash Point 11.5 %(V)
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss. p.a.
Linear Formula CH3COOC2H5
Vapor Pressure 97 hPa (20°C)
UN Number UN1173
Viscosity No data available
Beilstein 506104
Concentration ≥99.50%
Percent Purity ≥99.50%
Grade ACS
Chemical Name or Material Ethyl acetate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethyl acetate
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.