missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, 97%, Thermo Scientific™

Click to view available options
Quantity:
1g
10g
25g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
71083-06-2
C12H10FNO3
235.214
MFCD00052242
MPUYCZQHTGRPNE-UHFFFAOYSA-N
ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate, ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate, 3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester, 8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester, 8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester, pubchem19599, maybridge1_000540, acmc-209nc3
707160
ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F
This item is not returnable. View return policy
71083-06-2
C12H10FNO3
235.214
MFCD00052242
MPUYCZQHTGRPNE-UHFFFAOYSA-N
ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate, ethyl 8-fluoro-4-oxo-1,4-dihydroquinoline-3-carboxylate, ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate, 3-quinolinecarboxylicacid, 8-fluoro-1,4-dihydro-4-oxo-, ethyl ester, 8-fluoro-4-hydroxy-quinoline-3-carboxylic acid ethyl ester, 8-fluoro-4-hydroxyquinoline-3-carboxylic acid ethyl ester, 3-quinolinecarboxylic acid, 8-fluoro-4-hydroxy-, ethyl ester, pubchem19599, maybridge1_000540, acmc-209nc3
707160
ethyl 8-fluoro-4-oxo-1H-quinoline-3-carboxylate
CCOC(=O)C1=CNC2=C(C1=O)C=CC=C2F
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1g
Formula Weight 235.21
Percent Purity 97%
Chemical Name or Material Ethyl 8-fluoro-4-hydroxy-3-quinolinecarboxylate