missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-chloroimidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15469005
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15469005 1 g 1g
15479005 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15469005

Brand: Thermo Scientific Alfa Aesar H35348.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1204501-38-1
Molecular Formula C10H9ClN2O2
Molecular Weight (g/mol) 224.64
MDL Number MFCD11505035
InChI Key ZGSKNRFGZMYUGO-UHFFFAOYSA-N
Synonym ethyl 7-chloroimidazo 1,2-a pyridine-2-carboxylate, 7-chloro-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, ethyl 7-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylate, imidazo 1,2-a pyridine-2-carboxylic acid,7-chloro-,ethyl ester
PubChem CID 51063903
IUPAC Name ethyl 7-chloroimidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=CN2C=CC(Cl)=CC2=N1

Specifications

Melting Point 177°C to 179°C
Quantity 1 g
Formula Weight 224.65
Percent Purity 95%
Chemical Name or Material Ethyl 7-chloroimidazo[1,2-a]pyridine-2-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.