missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-bromoheptanoate, TRC

Product Code. 30386632
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386632 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30386632 Supplier Toronto Research Chemicals Supplier No. TRCE9007235G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material ethyl 7-bromoheptanoate
CAS 29823-18-5
Molecular Formula C9 H17 Br O2
Quantity 5 g
Synonym 7-Bromoheptanoic acid ethyl ester, Ethyl 7-bromoheptanoate, Tianeptine Imp. A (EP), Heptanoic acid, 7-bromo-, ethyl ester (6CI, 8CI, 9CI, ACI)
InChI Formula InChI=1S/C9H17BrO2/c1-2-12-9(11)7-5-3-4-6-8-10/h2-8H2,1H3
SMILES CCOC(=O)CCCCCCBr
IUPAC Name ethyl 7-bromoheptanoate
Molecular Weight (g/mol) 237.13
Formula Weight 236.0412
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.