missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 7-Aminothieno[2,3-b]pyrazine-6-carboxylate, TRC

Product Code. 30383469
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383469 500 mg
30377052 100 mg
30382770 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383469 Supplier Toronto Research Chemicals Supplier No. TRCE899665500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56881-21-1
Molecular Formula C9H9N3O2S
Molecular Weight (g/mol) 223.25
SMILES CCOC(=O)c1sc2nccnc2c1N

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C9H9N3O2S/c1-2-14-9(13)7-5(10)6-8(15-7)12-4-3-11-6/h3-4H,2,10H2,1H3
Chemical Name or Material Ethyl 7-Aminothieno[2,3-b]pyrazine-6-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.