missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-chloropyridazine-3-carboxylate, 95%

Product Code. 10217517 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10217517 1 g Glass bottle 1g
10710350 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217517 Supplier Thermo Scientific Supplier No. 431110010
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
This item is not returnable. View return policy

CAS: 75680-92-1 | C7H7ClN2O2 | 186.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75680-92-1
Molecular Formula C7H7ClN2O2
Molecular Weight (g/mol) 186.6
MDL Number MFCD09908168
InChI Key GVSVPKDEHFOXSW-UHFFFAOYSA-N
Synonym ethyl 6-chloro-3-pyridazinecarboxylate, 6-chloro-pyridazine-3-carboxylic acid ethyl ester, 3-pyridazinecarboxylic acid, 6-chloro-, ethyl ester, 6-chloropyridazine-3-carboxylic acid ethyl ester, 6-chloro-3-pyridazinecarboxylic acid ethyl ester, pubchem17694, acmc-1bd9t, 6-chloro-pyridazine-3-carboxylicacidethylester, ksc641q6n, ethyl 3-chloropyridazine-6-carboxylate
PubChem CID 10352425
IUPAC Name ethyl 6-chloropyridazine-3-carboxylate
SMILES CCOC(=O)C1=NN=C(C=C1)Cl

Specifications

Melting Point 149.0°C to 152.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186.6
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material Ethyl 6-chloropyridazine-3-carboxylate
compliance-icons
Product Identifier
  • Ethyl 6-chloropyridazine-3-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.