missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-(Chloromethyl)pyridine-2-carboxylate, TRC

Product Code. 30383015
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383015 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383015 Supplier Toronto Research Chemicals Supplier No. TRCE903613250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 6-(chloromethyl)pyridine-2-carboxylate
CAS 49668-99-7
Molecular Formula C9 H10 Cl N O2
Quantity 250 mg
Synonym 6-(Chloromethyl)pyridine-2-carboxylic Acid Ethyl Ester
InChI Formula InChI=1S/C9H10ClNO2/c1-2-13-9(12)8-5-3-4-7(6-10)11-8/h3-5H,2,6H2,1H3
SMILES O=C(OCC)C=1N=C(C=CC1)CCl
IUPAC Name ethyl 6-(chloromethyl)pyridine-2-carboxylate
Molecular Weight (g/mol) 199.6342
Formula Weight 199.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.