missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate, 95%, Thermo Scientific™

Product Code. 15459055
Change view
Klicka för att se tillgängliga alternativ
Quantity:
250 mg
1 g
5 g
Förpackningsstorlek:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
15459055 250 mg 250mg
15439055 1 g 1g
15449055 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 15459055 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer H35908.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 67625-37-0
Molecular Formula C10H9BrN2O2
Molecular Weight (g/mol) 269.098
MDL Number MFCD06659034
InChI Key MWDCKDQQAFZDOH-UHFFFAOYSA-N
PubChem CID 7141939
IUPAC Name ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate
SMILES CCOC(=O)C1=CN2C=C(C=CC2=N1)Br

Specifikationer

Melting Point 69°C to 72°C
Quantity 250 mg
Synonym ethyl 6-bromoimidazo 1,2-a pyridine-2-carboxylate, 6-bromo-imidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, imidazo 1,2-a pyridine-2-carboxylic acid, 6-bromo-, ethyl ester, 6-bromoimidazo 1,2-a pyridine-2-carboxylic acid ethyl ester, 6-bromo-2-ethoxycarbonyl imidazo 1,2-a pyridine, ethyl6-bromoimidazo 1,2-a pyridine-2-carboxylate, ethyl 6-bromoimidazo 1,2-a picolinate, 6-bromo-2-ethoxycarbonyl-imidazo 1,2-a pyridine, ethyl6-bromoh-imidazo 1,2-a pyridine-2-carboxylate, 6-bromoimidazo 1,2-a pyridine-2-carboxylicacidethylester
Formula Weight 269.1
Percent Purity 95%
Chemical Name or Material Ethyl 6-bromoimidazo[1,2-a]pyridine-2-carboxylate
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.