missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromohexanoate, 98%

Product Code. 10343722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10343722 100 g Glass bottle 100g
10670873 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10343722 Supplier Thermo Scientific Acros Supplier No. 305921000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25542-62-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00000270
InChI Key DXBULVYHTICWKT-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953
PubChem CID 117544
IUPAC Name ethyl 6-bromohexanoate
SMILES CCOC(=O)CCCCCBr

Specifications

Color White to Beige
Density 1.2540g/mL
Boiling Point 128°C to 130°C (16.0 mmHg)
Flash Point 58,3°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula Br(CH2)5CO2C2H5
Refractive Index 1.4580 to 1.46
Quantity 100 g
Specific Gravity 1.254
Formula Weight 223.11
Percent Purity 98%
Physical Form Powder or Granular Powder
Chemical Name or Material Ethyl 6-bromohexanoate, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.