missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromohexanoate, 97+%

Product Code. p-7024152
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468327 10 g 10g
11478327 50 g 50g
11488327 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468327 Supplier Thermo Scientific Alfa Aesar Supplier No. A18398.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25542-62-5 | C8H15BrO2 | 223.11 g/mol

Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-bromohexanoate is used in the preparation of various carnitine derivatives in a carnitine transporter study. It is also used for alkylation of pentane-2,4-dione.

Solubility
Soluble in water (water, 64.76 mg/L @ 25°C (est.)).

Notes
Store away from strong oxidizing agents, strong bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25542-62-5
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00000270
InChI Key DXBULVYHTICWKT-UHFFFAOYSA-N
Synonym 6-bromohexanoic acid ethyl ester, ethyl 6-bromocaproate, ethyl 6-bromocapronate, 6-bromocaproic acid ethyl ester, ethyl omega-bromocaproate, hexanoic acid, 6-bromo-, ethyl ester, ethyl .omega.-bromocaproate, 6-bromo-hexanoic acid ethyl ester, ethyl 6bromohexanoate, pubchem3953
PubChem CID 117544
IUPAC Name ethyl 6-bromohexanoate
SMILES CCOC(=O)CCCCCBr

Specifications

Density 1.254
Boiling Point 128°C to 130°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4595
Quantity 10 g
Beilstein 1756051
Solubility Information Soluble in water (water,64.76mg/L @ 25°C (est.)).
Formula Weight 223.12
Percent Purity ≥97%
Chemical Name or Material Ethyl 6-bromohexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.