missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-bromo-2-methylquinoline-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10144492
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10144492 1 g 1g
10381353 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10144492 Supplier Thermo Scientific Maybridge Supplier No. BTB00099DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 62482-30-8
Molecular Formula C13H12BrNO2
Molecular Weight (g/mol) 294.15
MDL Number MFCD00115248
InChI Key KOVIYCANWGLBHA-UHFFFAOYSA-N
Synonym maybridge1_000196, 2-methyl-6-bromoquinoline-4-carboxylic acid ethyl ester
PubChem CID 305135
IUPAC Name ethyl 6-bromo-2-methylquinoline-4-carboxylate
SMILES CCOC(=O)C1=C2C=C(Br)C=CC2=NC(C)=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 294.15
Percent Purity 95%
Chemical Name or Material Ethyl 6-bromo-2-methylquinoline-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.