missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 6-aminohexanoate, 98%

Product Code. 11489440
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489440 1 g 1g
11479440 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11489440 Supplier Thermo Scientific Alfa Aesar Supplier No. H50296.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 371-34-6 | C8H17NO2 | 159.23 g/mol

Ethyl 6-aminohexanoate is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 6-aminohexanoate is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-34-6
Molecular Formula C8H17NO2
Molecular Weight (g/mol) 159.23
MDL Number MFCD00233495
InChI Key NJNQDCIAOXIFTB-UHFFFAOYSA-N
Synonym 6-aminohexanoic acid ethyl ester, ethyl 6-aminocaproate, ethyl-6-aminohexanoate, acmc-20aky1, ethyl-epsilon-aminocaproate, 6-aminocaproic acid ethyl ester, 6-amino-hexanoic acid ethyl ester, hexanoic acid, 6-amino-, ethyl ester
PubChem CID 406972
IUPAC Name ethyl 6-aminohexanoate
SMILES CCOC(=O)CCCCCN

Specifications

Linear Formula H2N(CH2)5CO2CH2CH3
Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 159.23
Percent Purity 98%
Chemical Name or Material Ethyl 6-aminohexanoate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.