missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-Propyl-1H-pyrazole-3-carboxylate, TRC

Product Code. 30371177
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
missing translation for 'unitSize':
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30371177 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30371177 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCE925890250MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 5-Propyl-1H-pyrazole-3-carboxylate
CAS 92945-27-2
Molecular Formula C9H14N2O2
Quantity 250 mg
Synonym Ethyl 3-Propyl-1H-pyrazole-5-carboxylate, Ethyl 3-n-Propylpyrazole-5-carboxylate, Ethyl 5-n-Propyl-1H-pyrazole-3-carboxylate
InChI Formula InChI=1S/C9H14N2O2/c1-3-5-7-6-8(11-10-7)9(12)13-4-2/h6H,3-5H2,1-2H3,(H,10,11)
SMILES O=C(C1=NNC(CCC)=C1)OCC
IUPAC Name ethyl 5-propyl-1H-pyrazole-3-carboxylate
Molecular Weight (g/mol) 182.22
Formula Weight 182.11
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.