missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-methylisoxazole-4-carboxylate, 96%, Thermo Scientific™

Product Code. 11359435
Change view
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359435 10 g 10g
11349435 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359435 Supplier Thermo Scientific Alfa Aesar Supplier No. L18647.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51135-73-0
Molecular Formula C7H9NO3
Molecular Weight (g/mol) 155.153
MDL Number MFCD01631119
InChI Key KOMSQTMQKWSQDW-UHFFFAOYSA-N
Synonym ethyl 5-methylisoxazole-4-carboxylate, 5-methyl-4-isoxazolecarboxylate, ethyl 5-methyl-4-isoxazolecarboxylate, 5-methylisoxazole-4-carboxylic acid ethyl ester, acmc-20aky5, ethyl5-methylisoxazole-4-carboxylate, ethyl-5-methylisoxazole-4-carboxylate, ethyl 5-methylisoxazol-4-yl carboxylate, ethyl 5-m+c3117ethyl-4-isoxazolecarboxylate
PubChem CID 7009317
IUPAC Name ethyl 5-methyl-1,2-oxazole-4-carboxylate
SMILES CCOC(=O)C1=C(ON=C1)C

Specifications

Boiling Point 95°C to 97°C (15 mmHg)
Refractive Index 1.4597
Quantity 10 g
Beilstein 122097
Formula Weight 155.15
Percent Purity 96%
Chemical Name or Material Ethyl 5-methylisoxazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.