missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-hydroxyindole-2-carboxylate, 98+%

Product Code. 11350204
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11350204

Brand: Thermo Scientific Alfa Aesar L00774.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 24985-85-1 | C11H11NO3 | 205.213 g/mol

Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-hydroxyindole-2-carboxylate is used as an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.

Solubility
Insoluble in water.

Notes
Store between 2-8 C. Store in cool, dryplace in a well ventilated area. Store in a tightly sealed container. Protect from moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 24985-85-1
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.213
MDL Number MFCD00015458
InChI Key WANAXLMRGYGCPC-UHFFFAOYSA-N
Synonym ethyl 5-hydroxyindole-2-carboxylate, 5-hydroxy-1h-indole-2-carboxylic acid ethyl ester, 1h-indole-2-carboxylic acid, 5-hydroxy-, ethyl ester, 5-hydroxyindole-2-carboxylic acid ethyl ester, ethyl-5-hydroxy-1h-indol-2-carboxylat, pubchem15850, acmc-1cs3a, 5-hydroxy-1h-indole-2-carboxylicacidethylester, 2-carbethoxy-5-hydroxy-indole, ethyl 5-hydroxy-1h-2-indolecarboxylate
PubChem CID 90677
IUPAC Name ethyl 5-hydroxy-1H-indole-2-carboxylate
SMILES CCOC(=O)C1=CC2=C(N1)C=CC(=C2)O

Specifications

Melting Point 150°C to 151°C
Quantity 1 g
Beilstein 476925
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 205.21
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-hydroxyindole-2-carboxylate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.