missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromovalerate, 98%

Product Code. 11413964
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11413964 500 g 500g
11493954 25 g 25g
11403964 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11413964 Supplier Thermo Scientific Alfa Aesar Supplier No. A11952.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14660-52-7 | C7H13BrO2 | 209.08 g/mol

Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-bromovalerate is employed in the preparation of S(γ-Carboxypropyl)-DL-homocysteine and S(δ-Carboxybutyl)-DL-homocysteine. It is also used in the preparation of ethyl 5-azidovalerate.

Solubility
Not miscible or difficult to mix in water.

Notes
Light Sensitive. Keep away from oxidizing agents. Store in dark.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14660-52-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00000266
InChI Key AFRWBGJRWRHQOV-UHFFFAOYSA-N
PubChem CID 84580
IUPAC Name ethyl 5-bromopentanoate
SMILES CCOC(=O)CCCCBr

Specifications

Density 1.306
Boiling Point 227°C
Flash Point 103°C (217°F)
Linear Formula Br(CH2)4CO2CH2CH3
Refractive Index 1.458
Quantity 500 g
Beilstein 1752023
Sensitivity Light sensitive
Synonym ethyl 5-bromovalerate, 5-bromopentanoic acid ethyl ester, pentanoic acid, 5-bromo-, ethyl ester, 5-bromovaleric acid ethyl ester, ethyl omega-bromovalerate, ethyl5-bromovalerate, ethyl-5-bromovalerate, pubchem3955, ethyl-5-bromo-valerate, ethyl-5-bromopentanoate
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 209.09
Percent Purity 98%
Chemical Name or Material Ethyl 5-bromovalerate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.