missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl(5-bromobenzofuran)-2-carboxylate, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Synonym | ethyl 5-bromobenzofuran-2-carboxylate, buttpark 34\07-75, 5-bromo-benzofuran-2-carboxylic acid ethyl ester, ethylbromobenzofurancarboxylate, ethyl5-bromobenzofuran-2-carboxylate, ethyl 5-bromo-2-benzofurancarboxylate, ethyl 5-bromo-benzofuran-2-carboxylate, 5-bromo-2-ethoxycarbonyl-1-benzofuran |
Formula Weight | 269.09 |
Percent Purity | 95% |
Chemical Name or Material | Ethyl (5-bromobenzofuran)-2-carboxylate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'