missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-bromo-2-chlorobenzoate, 98+%, Thermo Scientific™

Product Code. 11444113
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444113 100 g 100g
11434113 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444113 Supplier Thermo Scientific Alfa Aesar Supplier No. A10334.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76008-73-6
Molecular Formula C9H8BrClO2
Molecular Weight (g/mol) 263.515
MDL Number MFCD00013635
InChI Key AMGWDYLEMSMUIO-UHFFFAOYSA-N
Synonym 5-bromo-2-chlorobenzoic acid ethyl ester, ethyl5-bromo-2-chlorobenzoate, benzoic acid, 5-bromo-2-chloro-, ethyl ester, pubchem3702, acmc-1bb9d, ethyl-5-bromo-2-chlorobenzoate, ethyl 5-bromo-2-chloro-benzoate, a-ethyl 5-bromo-2-chlorobenzoate, attercop-chm at115127, ethyl 5-bromo2-chlorobenzaldehyde
PubChem CID 144749
IUPAC Name ethyl 5-bromo-2-chlorobenzoate
SMILES CCOC(=O)C1=C(C=CC(=C1)Br)Cl

Specifications

Density 1.48
Boiling Point 156°C to 157°C (13 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.559
Quantity 100 g
Beilstein 6501991
Formula Weight 263.52
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-bromo-2-chlorobenzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.