missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-aminobenzo[b]furan-2-carboxylate, 97%, Thermo Scientific™

Product Code. 10302260 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10302260 5 g 5g
10119749 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10302260 Supplier Thermo Scientific Acros Supplier No. 437970050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 174775-48-5
Molecular Formula C11H11NO3
Molecular Weight (g/mol) 205.21
MDL Number MFCD08275091
InChI Key YFFLLDHEEWSHQG-UHFFFAOYSA-N
Synonym ethyl 5-aminobenzofuran-2-carboxylate, 2-benzofurancarboxylic acid, 5-amino-, ethyl ester, ethyl 5-aminobenzo b furan-2-carboxylate, 5-amino-2-ethoxycarbonyl-1-benzofuran, 5-aminobenzofuran-2-carboxylic acid ethyl ester, 5-amino-benzofuran-2-carboxylic acid ethyl ester, ethyl 5-aminobenzo d furan-2-carboxylate, pubchem23808, ethylaminobenzofurancarboxylate, ksc494o5f
PubChem CID 9837138
IUPAC Name ethyl 5-amino-1-benzofuran-2-carboxylate
SMILES CCOC(=O)C1=CC2=CC(N)=CC=C2O1

Specifications

Melting Point 56°C to 64°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 205.22
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Ethyl 5-aminobenzo[b]furan-2-carboxylate
compliance-icons
Product Identifier
  • Ethyl 5-aminobenzo¢b!furan-2-carboxylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.