missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-3-methylisothiazole-4-carboxylate, 97%, Thermo Scientific™

Catalog Number p-4510037
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
34859-65-9
C7H10N2O2S
186.229
ZDGBWOZFPKNORL-UHFFFAOYSA-N
2820511
ethyl 5-amino-3-methyl-1,2-thiazole-4-carboxylate
CCOC(=O)C1=C(SN=C1C)N
This item is not returnable. View return policy
34859-65-9
C7H10N2O2S
186.229
ZDGBWOZFPKNORL-UHFFFAOYSA-N
2820511
ethyl 5-amino-3-methyl-1,2-thiazole-4-carboxylate
CCOC(=O)C1=C(SN=C1C)N
Assay Percent Range 97%
Quantity 250 mg
Synonym ethyl 5-amino-3-methylisothiazole-4-carboxylate, ethyl5-amino-3-methylisothiazole-4-carboxylate, ethyl 5-amino-3-methyl-isothiazole 4-carboxylate, maybridge1_004847, 5-amino-3-methyl-isothiazole-4-carboxylate ethyl ester, 5-amino-4-ethoxycarbonyl-3-methylisothiazole, 4-isothiazolecarboxylic acid, 5-amino-3-methyl-, ethyl ester, 4-isothiazolecarboxylicacid, 5-amino-3-methyl-, ethyl ester
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'