missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-3-methylisothiazole-4-carboxylate, 97%, Thermo Scientific™

Product Code. 10295794
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10295794 250 mg 250mg
10765213 1 g 1g
10536134 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10295794 Supplier Thermo Scientific Maybridge Supplier No. KM04205CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 34859-65-9
Molecular Formula C7H10N2O2S
Molecular Weight (g/mol) 186.229
InChI Key ZDGBWOZFPKNORL-UHFFFAOYSA-N
PubChem CID 2820511
IUPAC Name ethyl 5-amino-3-methyl-1,2-thiazole-4-carboxylate
SMILES CCOC(=O)C1=C(SN=C1C)N

Specifications

Assay Percent Range 97%
Quantity 250 mg
Synonym ethyl 5-amino-3-methylisothiazole-4-carboxylate, ethyl5-amino-3-methylisothiazole-4-carboxylate, ethyl 5-amino-3-methyl-isothiazole 4-carboxylate, maybridge1_004847, 5-amino-3-methyl-isothiazole-4-carboxylate ethyl ester, 5-amino-4-ethoxycarbonyl-3-methylisothiazole, 4-isothiazolecarboxylic acid, 5-amino-3-methyl-, ethyl ester, 4-isothiazolecarboxylicacid, 5-amino-3-methyl-, ethyl ester
Formula Weight 186.23
Percent Purity 97%
Chemical Name or Material Ethyl 5-amino-3-methylisothiazole-4-carboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.