missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate, 97+%

Product Code. p-7028676
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11322029 1 g 1g
11332029 5 g 5g
11342029 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11322029 Supplier Thermo Scientific Alfa Aesar Supplier No. H26079.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 23286-70-6 | C7H11N3O2 | 169.184 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23286-70-6
Molecular Formula C7H11N3O2
Molecular Weight (g/mol) 169.184
MDL Number MFCD01210846
InChI Key WOCMIZZYXHVSPS-UHFFFAOYSA-N
Synonym ethyl 5-amino-3-methyl-1h-pyrazole-4-carboxylate, 5-amino-3-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, ethyl 5-amino-3-methylpyrazole-4-carboxylate, ethyl 3-amino-5-methyl-2h-pyrazole-4-carboxylate, acmc-1co96, 5-amino-3-methyl-1h-pyrazole-4-carboxylicacidethylester, ethyl 3-amino-5-methylpyrazole-4-carboxylate, 3-amino-5-methyl-1h-pyrazole-4-carboxylic acid ethyl ester, 3-methyl-5-amino-1h-pyrazole-4-carboxylic acid ethyl ester, pyrazole-4-carboxylic acid, 3-amino-5-methyl-, ethyl ester
PubChem CID 683559
IUPAC Name ethyl 3-amino-5-methyl-1H-pyrazole-4-carboxylate
SMILES CCOC(=O)C1=C(NN=C1N)C

Especificaciones

Melting Point 108°C
Quantity 1 g
Formula Weight 169.18
Percent Purity 98%
Chemical Name or Material Ethyl 5-amino-3-methyl-1H-pyrazole-4-carboxylate

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.