missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 98+%, Thermo Scientific Chemicals

Product Code. 11399979
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11399979 1 g 1g
11389979 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399979 Supplier Thermo Scientific Alfa Aesar Supplier No. H25762.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6440-02-4
Molecular Formula C5H7N3O2S
Molecular Weight (g/mol) 173.19
MDL Number MFCD00778317
InChI Key ZUGOHJWUTNKRMG-UHFFFAOYSA-N
Synonym ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate, 5-amino-1,2,3-thiadiazole-4-carboxylic acid ethyl ester, 1,2,3-thiadiazole-4-carboxylic acid, 5-amino-, ethyl ester, 5-amino-1,2,3 thiadiazole-4-carboxylic acid ethyl ester, acmc-20am6n, tos-bb-0200, zugohjwutnkrmg-uhfffaoysa, 5-amino-4-carbethoxy-1,2,3-thiadiazole, 5-amino-4-ethoxycarbonyl-1,2,3-thiadiazole, 1,2,3-thiadiazole-4-carboxylicacid,5-amino-,ethylester
PubChem CID 1831000
IUPAC Name ethyl 5-aminothiadiazole-4-carboxylate
SMILES CCOC(=O)C1=C(SN=N1)N

Specifications

Melting Point 123°C to 125°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 173.19
Percent Purity ≥98%
Chemical Name or Material Ethyl 5-amino-1,2,3-thiadiazole-4-carboxylate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.