missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 5,5-dimethyl-2,4-dioxohexanoate, 97%, Thermo Scientific™

Product Code. 10245284
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10245284 25 g 25g
10767783 1 g 1g
10149282 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10245284 Supplier Thermo Scientific Maybridge Supplier No. TL00361EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 13395-36-3
Molecular Formula C10H16O4
Molecular Weight (g/mol) 200.234
InChI Key NIMKIMUBJFWPTD-UHFFFAOYSA-N
Synonym ethyl trimethylacetopyruvate, hexanoic acid, 5,5-dimethyl-2,4-dioxo-, ethyl ester, 5,5-dimethyl-2,4-dioxo-hexanoic acid ethyl ester, 5,5-dimethyl-2,4-dioxohexanoic acid ethyl ester, ethyl pivaloylpyruvate, pivaloylpyruvic acid ethyl ester, 3-pivaloylpyruvic acid ethyl ester, ethyl5,5-dimethyl-2,4-dioxohexanoate, trimethylacetopyruvic acid ethyl ester, ethyl 5,5-dimethyl-2,4-dioxo-hexanoate
PubChem CID 83403
IUPAC Name ethyl 5,5-dimethyl-2,4-dioxohexanoate
SMILES CCOC(=O)C(=O)CC(=O)C(C)(C)C

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 200.23
Percent Purity 97%
Chemical Name or Material Ethyl 5, 5-dimethyl-2, 4-dioxohexanoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.