missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(trifluoromethyl)benzoate, 99%

Product Code. 15418939
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15418939 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15418939 Supplier Thermo Scientific Alfa Aesar Supplier No. L11139.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 583-02-8 | C10H9F3O2 | 218.175 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 583-02-8
Boiling Point 78°C to 80°C (5 mmHg)
Molecular Formula C10H9F3O2
Refractive Index 1.447
MDL Number MFCD00013563
Quantity 1 g
Beilstein 1968247
Synonym ethyl 4-trifluoromethyl benzoate, 4-trifluoromethyl benzoic acid ethyl ester, 4-trifluoromethylbenzoic acid, ethyl ester, pubchem16913, acmc-1cs5c, ethyl-p-trifluoromethylbenzoate, ethyl-4-trifluoromethyl benzoate, 4-trifluoromethylbenzoic acid ethyl ester
InChI Key ZQDADDSPMCHZPX-UHFFFAOYSA-N
SMILES CCOC(=O)C1=CC=C(C=C1)C(F)(F)F
IUPAC Name ethyl 4-(trifluoromethyl)benzoate
Molecular Weight (g/mol) 218.175
PubChem CID 521827
Formula Weight 218.18
Percent Purity 99%
Chemical Name or Material Ethyl 4-(trifluoromethyl)benzoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.