missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-(tert-Butyl)benzenesulfonate, TRC

Product Code. 30382205
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382205 500 mg 500mg
30381331 100 mg -
30387365 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382205 Supplier Toronto Research Chemicals Supplier No. TRCE701930500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 339370-19-3
Molecular Formula C12H18O3S
Molecular Weight (g/mol) 242.33
Synonym 4-(1,1-Dimethylethyl)-ethyl ester-benzenesulfonic Acid
SMILES CCOS(=O)(=O)c1ccc(cc1)C(C)(C)C

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H18O3S/c1-5-15-16(13,14)11-8-6-10(7-9-11)12(2,3)4/h6-9H,5H2,1-4H3
Formula Weight 242.0977
Chemical Name or Material Ethyl 4-(tert-Butyl)benzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.