missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-piperidone-3-carboxylate hydrochloride, 97%

Product Code. 11440083
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11440083 100 g 100g
11420083 5 g 5g
11430083 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11440083 Supplier Thermo Scientific Alfa Aesar Supplier No. A12131.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4644-61-5 | C8H14ClNO3 | 207.654 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4644-61-5
Molecular Formula C8H14ClNO3
Molecular Weight (g/mol) 207.654
MDL Number MFCD00043267
InChI Key RTYXWLMQWSFXNI-UHFFFAOYSA-N
Synonym ethyl 4-piperidone-3-carboxylate hydrochloride, ethyl 4-oxopiperidine-3-carboxylate hydrochloride, 3-carbethoxy-4-piperidone hydrochloride, ethyl 4-oxo-3-piperidinecarboxylate hydrochloride, 3-carbethoxy-4-piperidone hcl, 3-ethoxycarbonyl-4-piperidone hydrochloride, ethyl 4-piperidone-3-carboxylate hcl, ethyl 4-piperidone-3-carboxylate, hcl, 3-piperidinecarboxylic acid, 4-oxo-, ethyl ester, hydrochloride, 4-oxo-piperidine-3-carboxylic acid ethyl ester hydrochloride
PubChem CID 107315
IUPAC Name ethyl 4-oxopiperidine-3-carboxylate;hydrochloride
SMILES CCOC(=O)C1CNCCC1=O.Cl

Specifications

Melting Point ∼172°C (decomposition)
Quantity 100 g
Beilstein 3711279
Formula Weight 207.66
Percent Purity 97%
Chemical Name or Material Ethyl 4-piperidone-3-carboxylate hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.