Learn More
Ethyl 4-pentenoate, 98+%
Description
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Density | 0.9 |
| Boiling Point | 146°C |
| Flash Point | 37°C (98°F) |
| Refractive Index | 1.417 |
| Quantity | 5 g |
| UN Number | UN3272 |
| Beilstein | 1751643 |
| Solubility Information | Miscible with alcohol. Immiscible with water. |
| Formula Weight | 128.17 |
| Percent Purity | ≥98% |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.