missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 4-pentenoate, 98+%

Product Code. 11419623
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419623 100 g 100g
11458930 1 g 1g
11499613 5 g 5g
11409623 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11419623 Supplier Thermo Scientific Alfa Aesar Supplier No. A12422.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1968-40-7 | C7H12O2 | 128.171 g/mol

Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 4-pentenoate is found in leaf oil of Prunus phaeosticta phaeosticta from Taiwan. It is also used in the preparation of 5-iodomethyl-dihydro-furan-2-one by reacting with iodine.

Solubility
Miscible with alcohol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1968-40-7
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00036562
InChI Key PTVSRINJXWDIKP-UHFFFAOYSA-N
Synonym ethyl 4-pentenoate, 4-pentenoic acid ethyl ester, 4-pentenoic acid, ethyl ester, ethyl pent-4-en-1-oate, unii-210um0f8jp, 4-ethoxycarbonylbut-1-ene, pent-4-enoic acid ethyl ester, ethyl allylacetate, ethyl 4-pentenate, ethylpent-4-enoate
PubChem CID 74786
IUPAC Name ethyl pent-4-enoate
SMILES CCOC(=O)CCC=C

Specifications

Density 0.9
Boiling Point 146°C
Flash Point 37°C (98°F)
Refractive Index 1.417
Quantity 100 g
UN Number UN3272
Beilstein 1751643
Solubility Information Miscible with alcohol. Immiscible with water.
Formula Weight 128.17
Percent Purity ≥98%
Chemical Name or Material Ethyl 4-pentenoate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.